Rubicon™ Pro Grade Protective Eyewear, MCR Safety
Supplier: MCR Safety
T4110AF T4113AF T411R
89015-604EA 79.87 CAD
89015-604 89015-606 89015-608 89015-610 89015-612
Rubicon™ Pro Grade Protective Eyewear, MCR Safety
Safety Glasses
These glasses feature a curved, 180° single lens design with a close orbital seal for increased clarity and protection.
  • Lenses are scratch-resistant and filter out 99% of UV radiation
  • Platinum color metal temples with resilient spring hinges provide maximum flexibility
  • Soft, secure gel nosepiece and nonslip gel temple sleeves provide a comfortable fit for extended wear
  • Antifog lenses feature exclusive Duramass AF4® antifog lens coating
Certifications: Eyewear meets or exceeds ANSI Z87+ standards.
Ordering information: Each pair comes with a breakaway cord and a black satin carrying case that can be used as a cleaning cloth.