Histamine Free Base
Supplier: MilliporeSigma
3779-1GM 3779-5GM
CA80056-690EA 431.48 CAD
CA80056-690 CA80056-692
Histamine Free Base
Enzymes
A potent vasodilator found in normal tissues and blood
Activates nitric oxide synthase. Formed by the decarboxylation of L-histidine. Activation of histamine H1 receptors leads to Ca²⁺ mobilization, whereas histamine H2 receptor activation stimulates adenylate cyclase activity in neurons.