1400 Series Visitor Protective Eyewear, Encon® Safety Products
Supplier: Encon Safety Products
05148002
46500-071CS 1661.04 CAD
46500-071
1400 Series Visitor Protective Eyewear, Encon® Safety Products
Safety Glasses
Eyewear is flexible enough for comfort during extended use and strong enough to provide maximum impact protection.
  • One-piece spherical lens design offers 99% ultraviolet filtration
  • Equipped with polycarbonate side shields for high-impact absorption
Glasses feature a double pin hinge for added durability, and a universal nose bridge for a snug, secure fit.
Certifications: Meet ANSI Z87.1-2003 and CSA Z94.3-2002 protection standards. CE marked.