Upchurch Scientific® FEP Fluoropolymer Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
1520L 1548L 1521L 1521XL 1524L 1524XL 1650L 1650XL 1527L 1520XL 1548XL
21511-376EA 299.76 CAD
21511-376 53500-834 21511-378 53501-404 21001-032 21001-034 13500-012 13500-014 53500-830 53500-832 53500-836
Upchurch Scientific® FEP Fluoropolymer Tubing, IDEX Health & Science
Tubing Transfer Tubing
With virtually identical chemical resistance to PFA at a lower price, Upchurch Scientific® tubing made of DuPont® FEP fluoropolymer is great for general, low pressure applications.
  • The maximum recommended continuous use temperature for FEP tubing is 50 °C
Compared to PTFE, FEP (fluorinated ethylene propylene) tubing is held to tighter tolerances and has lower gas permeability. Much of this line of FEP tubing, even the color-tinted options—is translucent, making it possible to watch fluid flow. Using different colored tubing can help identify transferMore Product Information