Camco® Erado-Sol® Biological and Chemical Stain Remover, Cambridge
Supplier: Cambridge Diagnostic Products
207-16 207-32 207-G 307-8 307-16 307-32 307-G 307-2
CACM207EA 45.63 CAD
CACM207 CACM207C CA10853-876 CA75833-976 CA75833-978 CA75833-980 CA75833-982 CA75833-984
Camco® Erado-Sol® Biological and Chemical Stain Remover, Cambridge
Skincare Soaps and Skin Cleansers
Fast and effective stain remover for most medical, histological, and chemical stains including iodine, Wright stain, Lugol’s stain, and fresh blood
Removes stains from hands, plastic, glassware, and laboratory equipment. Simply rub in gently, rinse with water, and stain is removed in a matter of seconds. Stain remover is biodegradable and phosphate-free with a pH of 7–8.
Order Now
SPECIFICATIONS
Environmentally Preferable