Uvex Horizon™ Flip-Up Safety Eyewear, Honeywell Safety
Supplier: Honeywell Safety
uvex horizon™
S213 S213 S212
CA80086-570EA 60.38 CAD
80086-572 CA80086-570 CA80086-572
Uvex Horizon™ Flip-Up Safety Eyewear, Honeywell Safety
Safety Glasses
Flip-up style eyewear is ideal for applications requiring a shaded lens while working with high-intensity light sources and a traditional clear lens when the light-intensive task is complete.
  • Lightweight frame has sideshields for extra protection from flying particles
  • Clear lenses are made of impact-resistant polycarbonate
  • Feature a scratch-resistant hard coat for longer life
  • Block 99.9% of UVA and UVB radiation
  • Clear lens may also be replaced with prescription lenses
Flip-up lenses are available with two different lens coatings for various applications. The Infra-dura® 3.0 lens is ideal for use around welding sites and torch soldering. The Infra-dura® 5.0 block out even more infrared light and are ideal for use in welding, brazing, and cutting applications.
Certifications: ANSI Z87+