Uvex Genesis XC™ Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
S3301 S3301X S3306HS S3300 S3301 S3301X S3300 S3300HS S3301HS
14236-371EA 64.67 CAD
14236-371 14236-373 14236-377 14236-381 14236-383 14236-387 14236-389 14236-391 CA14236-362 CA14236-370 CA14236-372 CA14236-376 CA14236-380 CA14236-384 CA14236-386 CA14236-388 CA14236-390 CA14236-392 CA94023-918 14236-363 10790-628 10790-656
Uvex Genesis XC™ Protective Eyewear, Honeywell Safety
Safety Glasses
Wraparound safety eyewear offers extended protection from particles and debris with more than 180° of peripheral coverage.
  • Multi-Material Technology® (MMT) molding provides all-day comfort
  • Ergonomically designed frame relieves pressure points
  • Lenses and cushioned temples can be adjusted for a more personalized fit
Several lens tints are available to suit most work applications. These include SCT Spectrum Control Technology® lenses that absorb select wavelengths of radiant energy, and Infra-dura® shades for welding, brazing, and cutting. The Ultra-dura® lens coating provides superior scratch and chemical resistance. The Uvextreme® AF lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection. Dura-streme™ lens coating features a combination of Ultra-dura® on the exterior and Uvextreme® on the interior for excellent protection and longevity.
Certifications: ANSI Z87+ /CSA Z94.3