Blood Drawing Chairs, Labconco®
Supplier: Labconco
1132200 1132201 1120010 1120110 1120210
97007-598EA 5411.5 CAD
97007-598 97007-600 10766-666 10766-668 10766-670
Blood Drawing Chairs, Labconco®
Chairs
Chairs feature a durable, scratch-resistant seat mold, a welded frame with an elevated 25” chair height, and an ergonomically designed padded arm specifically contoured and angled at 10° to support the patient’s arm in a fully extended position for easier venipuncture
The padded arm equipped with a lift mechanism that assists the arm to the upright/vertical position, holds it stationary, and helps safely and gently lower the arm. The padded arm is constructed of molded urethane foam with an integral coating that cleans easily and withstands disinfection with alcohol or bleach. A Type 304 stainless steel footrest provides support for shorter and elderly patients, and provides easy access to and from the elevated seat.
The optional storage cabinet has a 35.6Wx51.4Dcm (14x20.25") slide-out, removable tray constructed of high-impact ABS plastic. The padded urethane cabinet top has a dished compartment for safe placement of test tubes and other blood drawing supplies. The double blood drawing chair features two chairs with a single shared storage cabinet at center. All chairs are glacier white, including frame, with charcoal arm and cabinet top. Chairs have a weight capacity of 204kg (450lbs.).
Bariatric blood drawing chairs feature a 96.5cm (38") wide upholstered seat, raised footrest, and optional storage cabinet. Padded upholstered armrests are adjustable in height and front/back position. Bariatric chairs have a weight capacity of 317.5kg (700lbs.).
The steel frame is coated with chemical-resistant powder epoxy. The wide chair frame and adjustable padded arm and seat accommodate large and small patients and have a weight capacity of 204kg (450lbs.)
Ordering information: Ten-year warranty (does not include normal wear of upholstery). Chairs are shipped fully assembled. Cabinets are shipped separately with legs and are easily attached with simple tools. Cabinets are interchangeable on the right or left side.