Uvex Skyper® Eyewear, Honeywell Safety
Supplier: Avantor
26902-681EA 45.9 CAD
26902-681 CA26902-680 CA26902-682
Uvex Skyper® Eyewear, Honeywell Safety
Safety Glasses
Wraparound safety glasses offer exceptional protection, comfort, and fit.
Stylish unilens features a molded-in nose bridge, side shields, and brow guard for complete coverage. Unique, adjustable Duoflex™ temples and adjustable lens inclination provide a custom fit. Impact-resistant, polycarbonate lens blocks 99.9% UV radiation. Available with Ultra-dura® hardcoat for scratch and chemical resistance, or Uvextreme® AF coating with antifog, anti-scratch, and antistatic properties.
Certifications: ANSI Z87.1 / CSA Z94.3
Ordering information: Replacement lenses available separately.