Uvex Genesis® Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
S3203 S3240 S3241X S3244 S3200 S3240 S3200HS S3201HS
14216-001EA 59.65 CAD
14216-001 14227-817 CA14216-000 CA14216-002 CA14227-815 CA14231-228 CA15001-568 CA15001-570 CA15001-576 CA15001-578 CA15001-582 CA15001-610 CA15001-612 CA15001-616 CA15001-620 CA15001-622 CA94023-916 15001-568 15001-610 10790-624 10790-626 CA14227-817
Uvex Genesis® Protective Eyewear, Honeywell Safety
Safety Glasses
Designed for comfort and compliance, Uvex Genesis® eyewear combines a lightweight, resilient multi-material frame with a soft elastomer brow guard for unsurpassed impact resistance.
  • Dual 9-base wraparound lens provides uncompromised peripheral vision and protection Elastomer brow-guard diffuses and deflects impact
  • Temple length and lens inclination are adjustable
  • Soft fingers conform to any nasal profile
  • Frames guaranteed for life
The dual 9-base wraparound polycarbonate lens provides uninterrupted peripheral coverage and visibility.
Clear lenses are appropriate for most indoor applications. Gold mirror, amber, and espresso lenses minimize the effects of sunlight and glare. The SCT-Reflect 50 lenses are designed for use in work applications with varying light conditions. This tint is a comfortable solution for workers who move between shaded or indoor locations and direct sunlight. SCT-Vermillion lenses are ideal for indoor applications requiring contrast, such as inspection. SCT-Blue lenses help counteract sodium vapor lighting. The Patriot® RWB frame features a red, white, and blue design with miniature U.S. flags at temples and 'United We Stand' inscribed on bottom right lens outside field of vision.
Soft, flexible 'fingers' adjust and conform to any nasal profile, minimizing slippage. A three-position ratchet lens inclination system completes the custom fit. A ventilation channel between frame and lens vents the eye socket area for outstanding moisture/fog control. Lens replacement is quick, easy, and economical. Ultra-dura® lens coating provides superior scratch and chemical resistance. Uvextreme® lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection. Dura-streme® lens coating features Ultra-dura® on the exterior and Uvextreme® on the interior for excellent protection and longevity.
Certifications: Eyewear meets military Vo ballistic test for impact and ANSI Z87+ and CSA Z94.3 standards.