Desiccator Cabinets, Acrylic, Plas-Labs™
Supplier: Plas-Labs
860-CG/EXP 860-CGA/EXP 861-CG/EXP 861-CGA/EXP 862-CG/EXP 862-CGA/EXP 863-CG/EXP 863-CGA/EXP 860-CG/6/EXP 860-CG/8/EXP 860CG/10/EXP 860CGA/6/EXP 860CGA/8/EXP 860CGA10/EXP
CA32940-030EA 3771.59 CAD
CA32940-030 CA32940-031 CA32940-032 CA32940-033 CA32940-034 CA32940-035 CA32940-036 CA32940-037 CA87000-742 CA87000-744 CA87000-746 CA87000-748 CA87000-750 CA87000-752
Desiccator Cabinets, Acrylic, Plas-Labs™
Desiccators Desiccator Cabinets
These nitrogen cabinets are designed to protect, store, or assist in transporting critical materials such as research specimens or moisture-sensitive devices that require a dry, contaminant-free environment
Mounted cabinets consist of several cubicles mounted on a stainless steel support cart with casters. These portable storage units remain stable during transport, making them safer than individually stacked cubicles.
Each cubicle has two nylon nipples to allow circulation of N2 gas, which prevents moisture from penetrating the molecular structure of the stored materials. The gas nipples have exterior 90° hose barb fittings that accept 0.250–0.375" I.D. tubing. Each cubicle features an airtight, gasketed door and a built-in dial hygrometer that measures relative humidity within the chamber. Interchangeable acrylic shelves have perforated slots for optimum circulation. Amber-tinted units help protect light-sensitive materials from harmful UV rays. Individual cubicles are stackable, with nonslip, grip-tight feet.
Ordering information: For custom configurations, contact your Avantor sales representative.