RheFlex® PEEK™ Fittings, Rheodyne®, IDEX Health & Science
Supplier: Rheodyne
6000-054 6000-055 6000-051
50806-930PK 126.8 CAD
50806-930 50806-932 50807-520
RheFlex® PEEK™ Fittings, Rheodyne®, IDEX Health & Science
Fittings and Connectors
RheFlex® fittings provide inert nonmetal connections to 10-32 internal thread ports, including all Rheodyne® injectors, valves, and sample loops
Nuts and ferrules do not bind to each other during tightening and are reusable. The standard fitting can be wrench or finger tightened, and the short fitting is wrench tightened only.
The 1/16" fitting will hold PEEK, stainless steel, and titanium tubing to 5000psi.
Ordering information: Set contains five nuts and five ferrules.