Stainless Steel Desiccating Cabinets, Boekel Scientific
Supplier: Boekel
1340 1342
24980-005EA 3438.4 CAD
24980-005 24980-107
Stainless Steel Desiccating Cabinets, Boekel Scientific
Desiccators Desiccator Cabinets
Completely airtight and moisture-proof
Includes two shelves (choice of Flexboard® material or stainless steel), 20.3Wx24.8Lcm (8x93/4"), with twelve 2.22cm (7/8") holes. Stainless steel desiccant tray, 20.3Wx22.9Lx1.6Dcm (8x9x5/8"), holds approximately 454g (1lb.) desiccant.
“Sealtight” door made of highly polished 18-8 stainless steel, with three sides of 4.75mm (3/16") glass. Manual relief valve normalizes pressure in cabinet before opening door. Indentations on top surface for stacking and storing.