Polysafe® / Polysafe® Plus Eyewear, Honeywell Safety
Supplier: Honeywell Safety
Polysafe® Plus Polysafe®
11180031 11180029 11180037 11180029
CA56611-447CS 413.83 CAD
CA56611-447 CA56611-448 56611-423 CA56611-423
Polysafe® / Polysafe® Plus Eyewear, Honeywell Safety
Safety Glasses
Eyewear provides a wide distortion-free field of vision, adjustable temples, and indirect ventilation to prevent fogging.
  • All styles feature clear lenses
  • Can be worn over most prescription eyewear (OTG compatible)
Wraparound polycarbonate lens incorporates side shields and a contoured nose bridge for added comfort. Polysafe® Plus styles feature a scratch-resistant coating on both sides of the lens for longer life and reduced glare.
Certifications: Meets ANSI Z87+ and CSA Z94.3 standards.