Norton 180® Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
T18000
CA89032-860EA 6.84 CAD
CA89032-860
Norton 180® Protective Eyewear, Honeywell Safety
Safety Glasses
Wraparound styling combines maximum protection with on-the-job comfort.
  • Eyewear is 100% lightweight polycarbonate with a clear
  • Scratch-resistant
  • One-piece wraparound lens
Vented side shield temples reduce heat buildup and lens fogging. Frosted brow reduces glare from overhead lighting. Four-post hinge system increases impact resistance by dispersing impact among additional contact points.
Certifications: ANSI Z87.1 / CSA Z94.3