Uvex Ultra-spec® Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety Products
S0112C S0250X S0300 S300CS
89096-706EA 43.67 CAD
89096-706 CA13500-856 CA13500-884 CA13500-886 CA14222-870 CA26902-472 CA33002-324
Uvex Ultra-spec® Protective Eyewear, Honeywell Safety
Safety Glasses
Economical glasses feature a 100% polycarbonate wraparound unilens design.
  • Features 100% polycarbonate wraparound unilens design
  • Side shields add extra protection without compromising visibility
  • Clear lenses and frames
  • The Ultra-dura® lens coating provides superior scratch and chemical resistance
  • Uvextreme® lens coating provides anti-fog, anti-scratch, anti-static, and ultaviolet protection
Uvex Ultra-spec® 2000 glasses feature an adjustable temple length. Uvex Ultra-spec® 2001 OTG glasses fit over prescription lenses and feature adjustable Duoflex™More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable