Spill Containment Trays, Scientific Plastics
Supplier: Scientific Plastics
T521916-101 T522219-101 T523419-101 T524619-101
89014-036EA 364.33 CAD
89014-036 89014-038 89014-040 89014-042
Spill Containment Trays, Scientific Plastics
Spill Control Materials
One-piece, rotationally-molded polyolefin trays protect cabinets and shelving from acid spills
Corners are rounded for easy cleaning. Walls are 4.8mm (3/16") thick for rigidity. Dishwasher safe. Depth: 2.4cm (1"). Color: natural.