Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
Supplier: Kemtech America
F501005 F501010 F501025
60004-218EA 35.21 CAD
60004-218 60004-220 60004-222
Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
Flasks Erlenmeyer Flasks
Erlenmeyer threaded flask for microscale applications.
  • Heavy wall borosilicate construction
  • Threaded upper joint
  • Open cap included
Supplied with a nylon compression cap, Viton® O-ring, and PTFE-lined silicone septum.