Synthware Flask, Pear-Shaped, Threaded, Microscale, Kemtech America
Supplier: Kemtech America
F321005 F321010 F321025
60004-188EA 46.94 CAD
60004-188 60004-190 60004-192
Synthware Flask, Pear-Shaped, Threaded, Microscale, Kemtech America
Flasks Distillation Flasks
Pear-shaped threaded flask for microscale applications.
  • Heavy wall borosilicate construction
  • Threaded joint
  • Compression cap included
Supplied with a nylon compression cap, Viton® O-ring, and PTFE-lined silicone septum.