VWR® Spilltray™ Tray and Drying Rack
Supplier: VWR
VWR® Spilltray™
3450
46610-022EA 67.67 CAD
46610-022
VWR® Spilltray™ Tray and Drying Rack
Spill Control Materials
Chemical-resistant, maintenance-free, polyethylene containment tray eliminates spills and holds 1.25L (42.3 oz.) of spilled liquid.
  • Safely contains chemical and liquid spills while pipetting, measuring, or mixing
  • Chemical-resistant, dishwasher-safe polyethylene tray holds 1.25L (42.3 oz.) of liquid
  • Low-profile design is perfect for drying glassware and delicate items
  • Plastic grid protects against breakage and scratches
  • Cuts drying time in half by permitting air circulation
Ideal for use with workstations or fume hoods to safely handle chemicals and liquids while pipetting, measuring, or mixing.
The low-profile design is perfect for drying glassware and delicate items. The white plastic grid cushions and protects against breakage and scratches, and cuts drying time in half by permitting air circulation. Entire unit is dishwasher safe.