Ultra C18 (USP L1) 3 and 5 µm Columns, Restek
Supplier: Restek
9174315 9174335 9174555 9174564 9174355 9174512 9174515 9174525 9174575 9174312 9174332 9174352 9174514 9174522 9174532 9174535 9174552 9174562 9174565 9174572 9174365
89047-714EA 1391.23 CAD
89047-714 89047-722 89047-764 89047-772 89047-730 89047-734 89047-740 89047-748 89047-782 89047-710 89047-718 89047-726 89047-738 89047-744 89047-752 89047-756 89047-760 89047-768 89047-774 89047-778 10059-742
Ultra C18 (USP L1) 3 and 5 µm Columns, Restek
Chromatography Columns
The ideal column for carbonyls, fatty acids, glycerides, anilines, fat-soluble vitamins, phthalates, barbiturates, steroids, PTH amino acids, and other acids.
  • Excellent peak shape across a range of compounds
  • Suited for general-purpose reversed phase applications
  • Fully endcapped
The retentive, high-purity packing exhibits excellent peak shape for a wide range of compounds. Columns are best suited for general-purpose reversed phase applications.