Formalin Absorbent Pads, Azer Scientific
Supplier: Azer Scientific
95039-810 ES728 ES730
95039-810CS 610.44 CAD
95039-810 CA95039-810 CA95039-812
Formalin Absorbent Pads, Azer Scientific
Spill Control Materials
Super absorbent pads are designed to absorb and neutralize up to 900 cc of 10% neutral buffered formalin.
  • Chemically hazardous vapors eliminated
  • Effective on 900 cc of 10% neutral buffered formalin
  • Extra absorbent
Pads are specially treated for the destruction of harmful vapors, including formaldehyde, 10% NBF, and glutaraldehyde solutions.
The blue color of each pad makes an excellent background for grossing tissue.
Ordering information: Absorbent pads 95039-810 are packaged 55 sheets per roll, six rolls per case. Absorbent pads 95039-812 are packaged 30 pads per case.