Nunc® OmniTray Single-Well Plates with Lids, Thermo Scientific
Supplier: Thermo Fisher Scientific
264728 165218 242811 140156
CA46600-638CS 884.18 CAD
CA46600-638 CA62409-590 CA62409-600 CA73520-160
Nunc® OmniTray Single-Well Plates with Lids, Thermo Scientific
Microplates HTS Microplates
Designed for many applications, including high throughput screening
These polystyrene, single-well plates have the same footprint as a 96-well plate. They also hold a 11.4x7.3cm (41/2x27/8") nylon membrane used for dot blotting bacterial clones. Lid protects cells from contamination while providing a means for cell respiration during incubation and/or storage. Radiation sterilized.
Nunclon™Δ certified surface ensures cell attachment and growth. MaxiSorp™ surface is ideal for arraying molecules with mixed hydrophilic/hydrophobic domains, such as proteins, directly onto the plate. Plate featuring an enhanced rectangular area is ideal for automatic picking and viewing systems.
Plates are useful for screening DNA libraries in high volume laboratories using robotics. They are also suitable for manual plating and screening. Compatible with Beckman Coulter®, Biomek®, Hybaid, Orca, Tecan® Trak, and Zymark® robotic systems.
Ordering information: Plates packaged 10 sleeves per case.