Synthware Valve, High Vacuum, Metering, PTFE-Protected, Low Hold Up Sidearms
Supplier: Kemtech America
S680004 S680008 S680012
60004-062EA 155.39 CAD
60004-062 60004-064 60004-066
Synthware Valve, High Vacuum, Metering, PTFE-Protected, Low Hold Up Sidearms
Valves Vacuum Valves
High vacuum metering valve with two 9 mm O.D
  • Hi vacuum valves
  • Heavy wall borosilicate construction
  • Teflon® protected
sidearms bent to minimize hold up of liquids. The Viton® O-rings on the PTFE plug shaft are protected with precision PTFE wipers. The tip of the plug has an integral O-ring to make the primary seal. This design provides an excellent vacuum-tight seal with PTFE as the only material exposed to chemicals.