Synthware Valve, High Vacuum, PTFE-Protected, 180° Sidearms, Kemtech America
Supplier: Kemtech America
S620004 S620008 S620012
60004-026EA 127.69 CAD
60004-026 60004-028 60004-030
Synthware Valve, High Vacuum, PTFE-Protected, 180° Sidearms, Kemtech America
Valves Vacuum Valves
High vacuum valve with two 9 mm O.D.
  • Hi Vacuum valves
  • Heavy wall Borosilicate construction
  • Teflon® Protected
sidearms sealed to the barrel at a 180° angle. The Viton® O-rings on the PTFE plug shaft are protected with precision PTFE wipers. The tip of the plug has an integral O-ring to make the primary seal. This design provides an excellent vacuum-tight seal with PTFE as the only material exposed to chemicals.