Sterilization Cabinets, Kerkau
Supplier: Kerkau
K-50SS H75SS K-50 F100SS H-75 F-100
53497-124EA 4106.14 CAD
53497-124 53497-122 53497-128 53497-126 53497-120 53497-130
Sterilization Cabinets, Kerkau
Sterilizers UV Sterilisers
These cabinets provide reliable infection control where workers share protective eyewear and headsets.
  • Durable, Heavy-Gauge Steel Construction
  • Ultraviolet Sanitation Processes
  • Safe and Easy to Use
  • Electrical Requirement: 110 V
  • Fifteen-Minute Timer for Controlled Sterilization Provided
The ultraviolet sanitation processes are lethal to virtually 100% of microorganisms
Large Cabinets with Reflectors have a capacity of 40 glasses and goggles or 20 headsets and are made of 24 gauge steel. Their aluminum reflectors increase ultraviolet light effectiveness. 24 gauge stainless steel Economical Cabinets have a capacity of 30 glasses and goggles. 22 gauge steel Cabinets with Reflectors have a capacity of 40 glasses and goggles. Aluminum reflectors increase ultraviolet light effectiveness.
Safe, easy-to-use steel cabinets have no messy dripping or drying and are available with painted or stainless steel exterior. They have an electrical requirement of 110 V.
Ordering information: All cabinets feature a 15-minute timer for controlled sterilization, safety door switch, key lock, long-life ultraviolet bulbs, and a heavy-duty 2.44 m (8') grounded neoprene power cord.