Centurion™ 454 Series Splash Safety Goggles, 3M™ Company
Supplier: 3M
Centurion™
40305-00000-10 40304-00000-10 40305-00000-10
CA33001-603CS 191.41 CAD
33001-601 33001-603 CA33001-603
Centurion™ 454 Series Splash Safety Goggles, 3M™ Company
Goggles
Centurion™ safety splash goggle is a reliable, economical choice in protective eyewear. Stable, sturdy construction will protect wearer's eyes.
  • Wide-angle wraparound lens and frame for good peripheral vision
  • Indirect venting system to help minimize fogging
  • Accommodates most prescription eyewear
Includes wide nasal flare for added comfort and an indirect venting system to minimize fogging. All goggles feature a strap attachment system.
Certifications: Meets ANSI Z87.1-2010, CSA Z94.3-2007.