Sonifier® Cell Disrupters, Branson Ultrasonics
Supplier: Branson Ultrasonics
101-063-196 101-063-199 101-063-201 101-063-200R 101-063-590 101-063-588 101-063-198 101-063-592 101-063-591 101-063-589
CA33995-309EA 13356.62 CAD
CA33995-309 CA33995-312 CA33995-319 CA33995-320 CA33995-454 CA33995-594 CA33995-310 CA33995-452 CA33995-584 CA33995-588 CA33995-592
Sonifier® Cell Disrupters, Branson Ultrasonics
Homogenizers Ultrasonic Cell Disruptors
Sonifier® Cell Disrupters are designed for biological and liquid processing applications where precise control and parameter measurement are required.
  • Adjustable amplitude
  • LCD display
  • Separate stop and pause buttons
  • Five different language setup menus
Front panel features an 80-character LCD display of parameter settings, alarms, and messages, and a bargraph display of relative power while the unit is running
Analog units feature a 0–15 minute timer with "Hold" position, duty cycle control with pulser mode for applying pulses of energy at one pulse a second (pulse duration variable from 0.1 to 0.9 seconds) and a continuous "On" position. Meter indicates percentage of ultrasonic power at any output setting.Soundproof enclosure reduces noise level up to 20db when liquids are treated ultrasonically. Especially helpful when using a cell disrupter for extended periods.
Preset up to 20 different parameter setups. Separate front panel stop and pause buttons for experimental cycles. Select from five different language setup menus (English, German, Italian, French, or Spanish). Parallel printer interface and serial interface for RS-232. Printed report capability with date stamp and parameter information. Self-diagnostics check performed on power-up.
Certifications: CE compliant. Model 150 is also UL listed and CSA certified.
Accessories information: The Sound proof enclosure reduces noise level up to 20db when liquids are treated ultrasonically. Especially helpful when using a cell disrupter for extended periods.
Ordering information: All units supplied with power supply and converter. Units supplied with a tapped horn also come with a flat replaceable tip. Units supplied with a solid horn also come with a solid tip. For additional disrupter horns and microtips, see 33995-323 series.