Stainless Steel Gowning Benches, Advance Tabco®
Supplier: Advance Tabco
BKR-4 BKR-5 BKR-6 BKR-7 BKR-8
58400-056EA 5143.25 CAD
58400-056 58400-058 58400-060 58400-062 58400-064
Stainless Steel Gowning Benches, Advance Tabco®
Furniture Gowning Benches
Durable, corrosion-resistant gowning benches provide sturdy support for workers dressing for the cleanroom environment.
  • Rolled rim front and back for worker comfort during changing
Constructed of 16-gauge, Type 304 stainless steel. Seat is 30.5 cm (12") wide; bench is 45.7 cm (18") high. Bench’s unique 'rainbow' leg design and 2.5 cm (1") adjustable plastic bullet feet permit freestanding use.
Ordering information: Benches can also be adapted for more secure floor mounting by ordering optional stainless steel flange feet. Units can beMore Product Information