SP Bel-Art Disposable Lens Cleaning Station, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F24835-0000
52848-152CS 823.42 CAD
52848-152
SP Bel-Art Disposable Lens Cleaning Station, Bel-Art Products, a part of SP
Cleaning Agents Lens Cleaning Solutions
Designed for economy and convenience, this self-contained, disposable station combines cleaning solution and lens tissue.
  • Disposable station design
  • Delivered ready to use
  • Tissues and cleaning fluid provided
The dispenser box is constructed of coated paperboard and has openings for wall mounting.
Lenses and cleaner are conveniently encased in dispenser. Tissues measure 12.7×20.3 cm (5×8"), packaged 600 count. Cleaning fluid is supplied in a 236.6 ml (8 oz.) container. Assembly not required.
Dispenses lens tissue and non-silicone cleaning fluid to protect, polish, antistat, and anti-fog lenses.