Lens Cleaning Solution, C-Clear
Supplier: C-Clear
1064681 44 1064671
11200-000CS 197.96 CAD
11200-000 11200-002 89094-546
Lens Cleaning Solution, C-Clear
Cleaning Agents Lens Cleaning Solutions
C-Clear lens cleaning solution is specially designed for the industrial setting.
  • Contains no silicone
  • Effectively cleans glass, plastic and polycarbonate lenses
  • Used on sideshields, face shields, goggles, respirators and CRT screens
This environmentally safe fluid cuts through grease and oil while leaving an anti-fog/anti-static coating on the lenses.
Order Now
SPECIFICATIONS
Environmentally Preferable