KleenGuard™ V80 Monogoggle™ VPC safety goggles offer splash protection that will fit comfortably over larger prescription eye glasses.
- Clear lens with a bronze frame
- Universal size (one size fits most); adjustable headband buckles for a snug fit
- Lenses are oversized to comfortably fit over larger prescription eyeglasses
- Indirect ventilation protects from splashes and provides increased airflow to minimize condensation and fogging
- Wider nose piece to fit over most half mask respirators and hard coated
KleenGuard™ V80 monogoggle VPC safety goggles offer the same two-piece goggle frame as other trusted Monogoggle™ styles, but with a larger lens for enhanced fit and comfort. They also feature a wider nose piece to fit over most half mask respirators. Each pair features indirect ventilation along with two additional baffled ports for increased airflow and minimized fogging and condensation.
The headband buckles are located toward the rear of the goggles to limit peripheral vision interference. The lenses are clear, making them ideal for indoor use and maximum light transmission. The polycarbonate lenses are hardcoated for scratch resistance and longer life, and they provide 99.9% UVA/UVB/UVC protection.
Certifications: Meet the ANSI Z87.1+ standards for impact, with a D3 rating for droplet and splash protection.