U-Tube Aniline Point Apparatus, Koehler
Supplier: Koehler
K10090 K10091
37000-006EA 19529.8 CAD
37000-006 37000-008
U-Tube Aniline Point Apparatus, Koehler
Petroleum Testing Apparatus
Used to characterize pure hydrocarbons and to indicate the aromatic content of hydrocarbon mixtures according to IP2-56 Method D
The apparatus consists of a U-form aniline sample tube, 400mL borosilicate glass beaker, cover assembly with bath stirrer, sample stirrer, and cooling coil, 6V lamp with line cord, and drive motor. Unit includes thermometer ferrules and mounting hardware. Available Powertrol heaters provide variable stepless control of heating rate and a low voltage receptacle for the 6V lamp.
Suitable for samples having 6.5 or lighter ASTM D1500 color.
Certifications: Conforms to specifications: ASTM D611, IP 2, ISO 2977, DIN 51775, FTM 791-3601.