Premium Face-Fit® Cover Goggles, Oberon
Supplier: Oberon
Face-Fit®
7006 7000
89040-878EA 76.33 CAD
89040-878 33009-005
Premium Face-Fit® Cover Goggles, Oberon
Goggles
Goggles feature a flexible PVC frame, green tinted for reduced glare and side reflections.
  • Elastic headband
  • Available uncoated or with anti-fog coating
Full face flange allows a better fit, including seal around the nose. Goggles fit over most prescription eyewear. Indirect ventilation provides splash protection. Injection-molded 0.127 cm (0.050") thick polycarbonate lens allows a distortion-free view.
Certifications: Meet ANSI Z87.1-1989 standard.