Synthware Repurification Solvent Still, Kemtech America
Supplier: Kemtech America
S134250 S134500
60004-750EA 695.71 CAD
60004-750 60004-752
Synthware Repurification Solvent Still, Kemtech America
Distillation Components Distillation Heads
This grease-free solvent still is supplied with the head only.
  • Heavy wall borosilicate construction
  • Ground glass joints
Condenser and flasks are not included. The access port has a threaded [ST] 14/10 joint designed to accept a PTFE-lined silicone septum with a compression cap (supplied) . The threaded [ST] 14/10 joint also accept regular 14/20 glass or PTFE stoppers (not supplied).