KIMBLE® KIMAX® Pink Colorware Beakers, Low Form, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
14000P-400 14000P-600 14000P-150 14000P-250 14000P-1000 14000P-50 14000P-100
10017-910CS 164.78 CAD
10017-910 10017-912 10017-914 10017-916 10018-426 10018-430 10018-432
KIMBLE® KIMAX® Pink Colorware Beakers, Low Form, DWK Life Sciences
Beakers
Please join us to support breast cancer prevention, treatment, and research by purchasing Pink Colorware from DWK Life Sciences.
  • With spout
  • Manufactured from borosilicate glass 3.3
  • Use of colored beakers reduces the risk of cross contamination
  • Easily identify glassware between labs
  • All sizes feature double capacity scales with easy-to-read graduations
  • Durable, color, matte finish marking area for use with an ordinary pencil or marker
Choose from a wide variety of KIMBLE® KIMAX® Pink “Colorware”, including Low Form Griffin Beakers, Narrow Mouth Erlenmeyer Flasks, Class A Volumetric Flasks, Blood Typing Tubes, and GL45 caps for ease of identification in the lab.
We will donate 20% of the proceeds to the Breast Cancer Research Foundation, the highest rated breast cancer organization in the United States. BCRF is recognized as one of the most financially efficient nonprofits in the country, and we are proud to assist in their support of breast cancer research.
Complies with ASTM Specification E960, Type I requirements.