Accessories for Compresstome® Specimen Tube Kits for VF-310-0Z and VF-210-0Z
Supplier: PRECISIONARY INSTRUMENTS LLC
VFBHSKVM20BOS VFBHSK15.5BOS VFSPSK15.5BOS VFSPSKVM20BOS
77435-412EA 1868.7 CAD
77435-412 77435-402 77435-476 77435-490
Accessories for Compresstome® Specimen Tube Kits for VF-310-0Z and VF-210-0Z
Microtomes and Microtome Assemblies Microtomes
Compresstome tube kits, with or without a blade holder are designed for use with Compresstome VF-310-0Z or VF-210-0Z models.
  • Specimen tube kits for use with VF-310-0Z or VF-210-0Z models
  • Kits are available with or without a blade holder
  • Available in 15.5 or 20 mm diameters
  • Full one year warranty
Compresstome specimen tube kits with a blade holder include two specimen tubes (15.5 or 20 mm) with matching plungers, one chilling block, one buffer tray, five plastic cleanup razors, one blade holder, one small Allen wrench, and one small screw.
Order Now
SPECIFICATIONS
Environmentally Preferable