Accessories for Compresstome® Legacy Model Specimen Tube
Supplier: PRECISIONARY INSTRUMENTS LLC
VFSPSVM12.5NC VFSPSVM15.5NC
77435-492EA 504.42 CAD
77435-492 77435-496
Accessories for Compresstome® Legacy Model Specimen Tube
Microtomes and Microtome Assemblies Microtomes
Compresstome legacy specimen tubes are compatible with compresstome models VF-200, VF-200-0Z, VF-300, and VF-300-0Z.
  • Specimen tube with 12.5 or 15.5 mm diameter
  • Full one year warranty
A replacement 12.5 or 15.5 mm compresstome specimen tube.
Ordering information: Includes 1 specimen tube with matching plunger.