Compresstome® Vibratomes
Supplier: PRECISIONARY INSTRUMENTS LLC
VF2100ZVMBOS VF300VMNC VF5100Z
77435-372EA 39553.85 CAD
77435-372 77435-374 77435-376
Compresstome® Vibratomes
Microtomes and Microtome Assemblies Microtomes
Compresstome® VF-510-0Z, VF-300-0Z, or VF-210-0Z vibratomes.
  • Compresstome® VF-510-0Z is fully automated to cut slices in 1 µm increments and includes a 5 year warranty
  • Compresstome® VF-300-0Z is fully automated for sectioning fixed and live tissues and includes a 5 year warranty
  • Compresstome® VF-210-0Z is a vibrating microtome for cutting tissue slices with Auto Zero-Z technology
The VF-510-0Z is the newest Compresstome® model and is fully automated. This vibratome comes calibrated with Auto Zero-Z patented technology, which ensures healthier liveMore Product Information