Verispec® Certified Reference Material/Standard, KCl Conductivity Standards at 25°C, RICCA Chemical Company
Supplier: Ricca Chemical
RV588684-500A RV588750-500A RV588775-500A RV588843-500A RV588860-500A
RCRV588684500AEA 104.28 CAD
RCRV588684500A RCRV588750500A RCRV588775500A RCRV588843500A RCRV588860500A
Verispec® Certified Reference Material/Standard, KCl Conductivity Standards at 25°C, RICCA Chemical Company
Standards Conductivity Standards
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: Store at room temperature. The conductivity of these certified reference materials are strongly dependent on temperature. All calibrations with these certified reference materials should be conducted at 25°C.