Primate MD's™ Anxiolytic
Supplier: Bio-Serv
MD930-500 MD930-500
77391-950EA 675.5 CAD
77391-950 CA77391-950
Primate MD's™ Anxiolytic
Animal Supplies Animal Feed
Chewable, flavored, medicated tablet replaces more stressful dosing methods.
  • Tablet consistency ensures accurate dosing
  • Scored 5 gm tablet for flexible dose administration
  • Cold tableting process eliminates drug degradation
  • No heat or water used in processing
  • Highly palatable sugar-based formula with excellent drug masking capabilities
  • Reduces time, labor and stress compared to traditional dosing methods
  • MD's can be customized to meet specific requirements
Shelf life: 6 to 24 months as dictated by the drug expiration date.
Base meal: Sucrose, dextrose, mineral oil, artificial and natural flavors, calcium silicate, magnesium stearate