Synthware Vacuum Transfer Adapters, Kemtech America
Supplier: Kemtech America
A121420 A121922 A122440
60000-326EA 339.02 CAD
60000-326 60000-328 60000-330
Synthware Vacuum Transfer Adapters, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Use for the transfer of sensitive and hazardous solutions.
  • [ST] joint
  • PTFE stopcock
Features a 2mm PTFE stopcock.