Puritan® Wooden Applicator/Stirring Sticks, Puritan Medical Products
Supplier: Puritan Medical Products
807 25-8072W 805 807 805
CA10805-018CS 866.99 CAD
CA10805-018 CA82030-606 CA82030-614 CA82030-616 82030-606 10805-018 82030-614 87006-044
Puritan® Wooden Applicator/Stirring Sticks, Puritan Medical Products
Stirrers Stirring Rods
Wooden applicator sticks are made for general purpose functionality.
Choose a size and shape that works best for required applications, such as specimen transfer, mixing rods, and laboratory work. Sticks are manufactured from northern maine white birch, with straight cut ends.
Ordering information: Available in sterile or nonsterile formats. Sterile items are individually wrapped, with two sticks per package.