Synthware Flushing Adapters, Kemtech America
Supplier: Kemtech America
A761420 A762440
60000-310EA 218.13 CAD
60000-310 60000-314
Synthware Flushing Adapters, Kemtech America
Adapters Adapters, Joints with Tube Connectors
The flushing adapter is used to flush and fill attached vessels.
  • [ST] joint
  • PTFE stopcock
Features a 2 mm T-bore PTFE stopcock.