RingSeal™ Cryogenic Vials
Supplier: Globe Scientific
3034-2-RB 3034-4 3034-5 3034-5-RB 3032-2-RB 3032-5-RB 3033-CIB 3033-CIG 3033-CIR 3033-CIW 3034-1
77509-258CS 214.37 CAD
77509-258 77509-260 77509-262 77509-264 77509-236 77509-242 77509-244 77509-246 77509-248 77509-250 77509-254
RingSeal™ Cryogenic Vials
Vials
A good choice for long-term sample storage. Built with reliable O-ring seals, this exceptional range offers multiple variants to meet diverse research needs.
  • RingSeal™ Cryogenic Vials provide leak-proof protection up to 90 kPa, safeguarding against potential leaks or contamination
  • Reliable O-ring seals effectively protect samples within a wide temperature range, from −196 to 121 °C
  • Choose between round bottom and self-standing vials, based on specific requirements
  • Certified DNase-, RNase-, and Pyrogen-free vials ensure the integrity and cleanliness of valuable samples
  • Available internal and external thread options allow compatibility with a variety of laboratory processes
  • Large writing area aids in convenient sample identification and organization
  • Autoclavable vials ensure efficient sterilization while maintaining structural integrity
Globe Scientific’s RingSeal™ Cryogenic vials are available in both internal and external thread options, as well as self-standing and round bottom formats. Vials are offered in the most popular volumes.