Synthware Flow Control Adapters with Metering Stopcock, 90° Angle, Kemtech America
Supplier: Kemtech America
A721420 A721922 A722440
60000-294EA 198.37 CAD
60000-294 60000-296 60000-298
Synthware Flow Control Adapters with Metering Stopcock, 90° Angle, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Provides on/off control with the main valve and allows delivery of preset volumes of gases or liquids to reactions using the needle valve.
  • 90° angle bend
  • [ST] joint
  • Metering stopcock
Stopcock bore is 2 mm.