Synthware Pressure Equalizing Funnel with High Vacuum Valve, Kemtech America
Supplier: Kemtech America
F641425 F641460 F641960 F642460 F641125 F649125 F644125 F644250
60004-382EA 372.24 CAD
60004-382 60004-384 60004-386 60004-388 60004-390 60004-392 60004-394 60004-398
Synthware Pressure Equalizing Funnel with High Vacuum Valve, Kemtech America
Funnels Separating Funnels
Funnels feature a PTFE extended tip valve for semi-needle valve adjustment.
  • Heavy wall borosilicate construction
  • Ground glass joints
  • High vacuum valve Included
  • Graduated upper column
This pressure equalizing funnel is ideal for the controlled transfer of liquids in a sealed, air sensitive environment.