Protektive Pak® ESD Storage Containers, Desco Industries
Supplier: Protektive Pak
37512 37514 37517
30000-986EA 76.11 CAD
30000-986 30001-180 30001-280
Protektive Pak® ESD Storage Containers, Desco Industries
Boxes ESD-Safe Boxes
Provide ESD and physical protection for ESD-sensitive components and devices
The conductive plastic handles provide easier handling of the container, as well as a reliable path-to-ground. The steel wire reinforcing frame provides additional strength and durability.
Complete Faraday cage protection. Containers are made of durable fluted conductive cardboard and feature an impregnated liner. Buried shielding layer of <104ohms/sq. minimizes sloughing and rub-off contamination. Feature a static-dissipative surface of 107–109ohms/sq. These reusable containers are constructed with doubleMore Product Information