Puritan® Foam-Over-Cotton Tipped Swab, Wood Handle, Puritan Medical Products
Supplier: Puritan Medical Products
1806-WCF 1806-WCFB 1806-WCSFB 1806-WCF 1806-WCFB 1806-WCSFB
CA82030-590CS 777.76 CAD
CA82030-590 CA82030-592 CA82030-594 82030-590 82030-592 82030-594
Puritan® Foam-Over-Cotton Tipped Swab, Wood Handle, Puritan Medical Products
Swabs Swabs for Sampling
This absorbent foam tipped applicator is produced with reticulated 100 ppi medical grade foam over high quality USP grade cotton.
It is bonded securely to a rigid white birch shaft. For use in critical environments and as cleaning applicators for particle removal.