PurSwab® Rectangular Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
1805PFB-RECT 1805-PF-RECT 1805-PF-RECT 1805-PFBRECT
CA10806-023CS 1284 CAD
CA10806-023 CA10806-032 10806-032 10806-023
PurSwab® Rectangular Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
Lint-free, nonabrasive, shred-resistant swabs are ideal for precision cleaning
Made in the USA.
Flat, slightly squared 100ppi polyurethane foam tip is securely bonded to the polypropylene shaft.
Certifications: CE marked.